α-Methyl-2-(1-methylethoxy)-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234862-1G 234862-500MG 234862-5G 234862-25G
77467-158EA 415.09 USD
77467-158 77479-882 77490-186 77500-390
α-Methyl-2-(1-methylethoxy)-α-phenylbenzenemethanol ≥95%
α-Methyl-2-(1-methylethoxy)-α-phenylbenzenemethanol

Precaución: For research use only

Formula: C₁₇H₂₀O₂
MW: 256.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23422859
CAS Number: 1534028-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR