4-Bromo-3-fluoro-α-methyl-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234783-1G 234783-500MG 234783-5G 234783-25G
77467-154EA 415.09 USD
77467-154 77479-878 77490-182 77500-386
4-Bromo-3-fluoro-α-methyl-α-phenylbenzenemethanol ≥95%
4-Bromo-3-fluoro-α-methyl-α-phenylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₂BrFO
MW: 295.15 g/mol
Storage Temperature: Ambient
CAS Number: 1196396-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR