4-Bromo-α-methyl-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234752-1G 234752-500MG 234752-5G
77467-150EA 471.74 USD
77467-150 77479-874 77490-178
4-Bromo-α-methyl-α-phenylbenzenemethanol ≥95%
4-Bromo-α-methyl-α-phenylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₃BrO
MW: 277.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12068085
CAS Number: 15832-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR