Cyclopropyl(3-ethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

234595-1G 234595-500MG 234595-5G
77467-142EA 576.8 USD
77467-142 77479-866 77490-170
Cyclopropyl(3-ethoxyphenyl)methanone ≥95%
Cyclopropyl(3-ethoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20335955
CAS Number: 1342071-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR