1-[3,5-Dimethyl-4-(phenylmethoxy)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

234547-1G 234547-500MG 234547-5G
77467-138EA 666.41 USD
77467-138 77479-862 77490-166
1-[3,5-Dimethyl-4-(phenylmethoxy)phenyl]ethanone ≥95%
1-[3,5-Dimethyl-4-(phenylmethoxy)phenyl]ethanone

Precaución: For research use only

Formula: C₁₇H₁₈O₂
MW: 254.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870106
CAS Number: 168106-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR