[1,1'-Biphenyl]-4-yl(3-chlorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

234401-1G 234401-500MG 234401-5G
77467-134EA 594.31 USD
77467-134 77479-858 77490-162
[1,1'-Biphenyl]-4-yl(3-chlorophenyl)methanone ≥95%
[1,1'-Biphenyl]-4-yl(3-chlorophenyl)methanone

Precaución: For research use only

Formula: C₁₉H₁₃ClO
MW: 292.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01662304
CAS Number: 98257-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR