2-Chloro-3,4-difluoro-1-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

23408-1G 23408-500MG 23408-5G 23408-25G
77467-122EA 188.49 USD
77467-122 77479-846 77490-150 77500-370
2-Chloro-3,4-difluoro-1-(methoxymethoxy)benzene ≥97%
2-Chloro-3,4-difluoro-1-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇ClF₂O₂
MW: 208.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561297
CAS Number: 749230-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR