1-[3-(1-Methylethyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

234069-1G 234069-500MG 234069-5G
77467-118EA 668.47 USD
77467-118 77479-842 77490-146
1-[3-(1-Methylethyl)phenyl]-1-propanone ≥95%
1-[3-(1-Methylethyl)phenyl]-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617655
CAS Number: 1342156-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR