1-(4-Ethoxy-3,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

233617-1G 233617-500MG 233617-5G 233617-25G
77467-110EA 352.26 USD
77467-110 77479-834 77490-138 77500-364
1-(4-Ethoxy-3,5-dimethylphenyl)ethanone ≥95%
1-(4-Ethoxy-3,5-dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MW: 192.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10485790
CAS Number: 113361-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR