5-(1,3-Dioxolan-2-yl)-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

23237-1G 23237-500MG 23237-5G
77467-098EA 576.8 USD
77467-098 77479-822 77490-126
5-(1,3-Dioxolan-2-yl)-2-fluoropyridine ≥97%
5-(1,3-Dioxolan-2-yl)-2-fluoropyridine

Precaución: For research use only

Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD18250377
CAS Number: 1034467-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR