2-Chloro-5-(methoxymethoxy)isonicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

23076-1G 23076-500MG 23076-5G 23076-25G
77467-094EA 368.74 USD
77467-094 77479-818 77490-122 77500-352
2-Chloro-5-(methoxymethoxy)isonicotinaldehyde ≥95%
2-Chloro-5-(methoxymethoxy)isonicotinaldehyde

Precaución: For research use only

Formula: C₈H₈ClNO₃
MW: 201.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27942957
CAS Number: 1282516-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR