3-Bromo-2-fluoropyridine-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

23016-1G 23016-500MG 23016-5G
77467-090EA 1192.74 USD
77467-090 77479-814 77490-116
3-Bromo-2-fluoropyridine-4-carboxylic acid ≥95%
3-Bromo-2-fluoropyridine-4-carboxylic acid

Precaución: For research use only

Formula: C₆H₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185800
CAS Number: 1214360-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR