α-Ethyl-6-methoxy-2-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

229880-1G 229880-500MG 229880-5G 229880-25G
77467-086EA 397.58 USD
77467-086 77479-810 77490-112 77500-346
α-Ethyl-6-methoxy-2-pyridinemethanol ≥95%
α-Ethyl-6-methoxy-2-pyridinemethanol

Precaución: For research use only

Formula: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34869735
CAS Number: 1697904-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR