1,3-Dimethoxy-5-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229451-1G 229451-500MG 229451-5G 229451-25G
77467-082EA 415.09 USD
77467-082 77479-806 77490-108 77500-342
1,3-Dimethoxy-5-[(1-methylethyl)thio]benzene ≥95%
1,3-Dimethoxy-5-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₁H₁₆O₂S
MW: 212.31 g/mol
Storage Temperature: Ambient
CAS Number: 952006-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR