1-(1-Methylethoxy)-3-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229401-1G 229401-500MG 229401-5G 229401-25G
77467-074EA 415.09 USD
77467-074 77479-798 77490-100 77500-334
1-(1-Methylethoxy)-3-[(1-methylethyl)thio]benzene ≥95%
1-(1-Methylethoxy)-3-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
CAS Number: 79128-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR