1,3-Dimethyl-5-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229380-1G 229380-500MG 229380-5G 229380-25G
77467-070EA 415.09 USD
77467-070 77479-794 77490-096 77500-330
1,3-Dimethyl-5-[(1-methylethyl)thio]benzene ≥95%
1,3-Dimethyl-5-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₁H₁₆S
MW: 180.31 g/mol
Storage Temperature: Ambient
CAS Number: 2254571-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR