4-Fluoro-1-methoxy-2-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229356-1G 229356-500MG 229356-5G 229356-25G
77467-066EA 415.09 USD
77467-066 77479-790 77490-092 77500-326
4-Fluoro-1-methoxy-2-[(1-methylethyl)thio]benzene ≥95%
4-Fluoro-1-methoxy-2-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₀H₁₃FOS
MW: 200.27 g/mol
Storage Temperature: Ambient
CAS Number: 2254577-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR