1-Ethyl-4-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229344-1G 229344-500MG 229344-5G
77467-062EA 736.45 USD
77467-062 77479-786 77490-088
1-Ethyl-4-[(1-methylethyl)thio]benzene ≥95%
1-Ethyl-4-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₁H₁₆S
MW: 180.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621797
CAS Number: 158321-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR