2-(5-Bromo-2,3-difluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

22841-1G 22841-500MG 22841-5G 22841-25G
77467-054EA 422.3 USD
77467-054 77479-778 77490-078 77500-316
2-(5-Bromo-2,3-difluorophenyl)-1,3-dioxolane ≥95%
2-(5-Bromo-2,3-difluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977091
CAS Number: 2221812-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR