2-Fluoro-5-methoxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

22683-1G 22683-500MG 22683-5G 22683-25G
77467-050EA 149.35 USD
77467-050 77479-774 77490-074 77500-312
2-Fluoro-5-methoxy-4-methylpyridine ≥97%
2-Fluoro-5-methoxy-4-methylpyridine

Precaución: For research use only

Formula: C₇H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD16611213
CAS Number: 1227596-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR