5-Fluoro-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

22635-1G 22635-500MG 22635-5G 22635-25G
77467-046EA 244.11 USD
77467-046 77479-770 77490-070 77500-310
5-Fluoro-2-(methylthio)phenol ≥97%
5-Fluoro-2-(methylthio)phenol

Precaución: For research use only

Formula: C₇H₇FOS
MW: 158.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998980
CAS Number: 1243442-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR