1-(6-Bromopyridin-3-yl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

226261-1G 226261-500MG 226261-5G
77467-042EA 525.3 USD
77467-042 77479-766 77490-066
1-(6-Bromopyridin-3-yl)-2-methylpropan-1-one ≥95%
1-(6-Bromopyridin-3-yl)-2-methylpropan-1-one

Precaución: For research use only

Formula: C₉H₁₀BrNO
MW: 228.0858 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31631276
CAS Number: 801303-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR