3-Fluoro-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22622-1G 22622-500MG 22622-5G 22622-25G
77467-038EA 178.19 USD
77467-038 77479-764 77490-062 77500-304
3-Fluoro-4-(methylthio)benzaldehyde ≥97%
3-Fluoro-4-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₈H₇FOS
Storage Temperature: Ambient
MDL Number: MFCD18887991
CAS Number: 177756-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR