1-Bromo-2-chloro-3-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

226099-1G 226099-500MG 226099-5G 226099-25G
77467-034EA 131.84 USD
77467-034 77479-760 77490-058 77500-300
1-Bromo-2-chloro-3-(trifluoromethoxy)benzene ≥95%
1-Bromo-2-chloro-3-(trifluoromethoxy)benzene

Precaución: For research use only

Formula: C₇H₃BrClF₃O
MW: 275.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22580763
CAS Number: 1417568-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR