1-Iodo-2-methoxy-4,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

225897-1G 225897-500MG 225897-5G
77467-030EA 551.05 USD
77467-030 77479-756 77490-054
1-Iodo-2-methoxy-4,5-dimethylbenzene ≥95%
1-Iodo-2-methoxy-4,5-dimethylbenzene

Precaución: For research use only

Formula: C₉H₁₁IO
MW: 262.09 g/mol
Storage Temperature: Ambient
CAS Number: 50772-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR