4-Ethoxy-2-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

22421-1G 22421-500MG 22421-5G 22421-25G
77467-026EA 108.15 USD
77467-026 77479-752 77490-050 77500-292
4-Ethoxy-2-methylbenzaldehyde ≥95%
4-Ethoxy-2-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06247360
CAS Number: 89763-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR