2,3-Dichloro-4-(methylsulfonyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

223369-1G 223369-500MG 223369-5G 223369-25G
77467-018EA 415.09 USD
77467-018 77479-744 77490-042 77500-284
2,3-Dichloro-4-(methylsulfonyl)benzoic acid ≥95%
2,3-Dichloro-4-(methylsulfonyl)benzoic acid

Precaución: For research use only

Formula: C₈H₆Cl₂O₄S
MW: 269.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06654029
CAS Number: 105917-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR