5-Chloro-2-(methylsulfonyl)benzoic acid ≥96%

Supplier: AOBChem USA

Warning

223312-1G 223312-500MG 223312-5G
77467-014EA 673.62 USD
77467-014 77479-740 77490-038
5-Chloro-2-(methylsulfonyl)benzoic acid ≥96%
5-Chloro-2-(methylsulfonyl)benzoic acid

Precaución: For research use only

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25966599
CAS Number: 720689-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR