1,2-Dimethyl-4-(methylsulfonyl)benzene ≥96%

Supplier: AOBChem USA

Warning

223125-1G 223125-500MG 223125-5G 223125-25G
77467-010EA 101.97 USD
77467-010 77479-738 77490-032 77500-276
1,2-Dimethyl-4-(methylsulfonyl)benzene ≥96%
1,2-Dimethyl-4-(methylsulfonyl)benzene

Precaución: For research use only

Formula: C₉H₁₂O₂S
MW: 184.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789178
CAS Number: 38452-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR