3-Fluoro-5-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22242-1G 22242-500MG 22242-5G 22242-25G
77467-006EA 338.87 USD
77467-006 77479-734 77490-028 77500-272
3-Fluoro-5-isopropoxybenzaldehyde ≥97%
3-Fluoro-5-isopropoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28716973
CAS Number: 1289030-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR