4-Bromo-5-methoxy-1,3-benzodioxole ≥95%

Supplier: AOBChem USA

Warning

222126-1G 222126-500MG 222126-5G
77467-002EA 653.02 USD
77467-002 77479-730 77490-024
4-Bromo-5-methoxy-1,3-benzodioxole ≥95%
4-Bromo-5-methoxy-1,3-benzodioxole

Precaución: For research use only

Formula: C₈H₇BrO₃
MW: 231.04 g/mol
Storage Temperature: Ambient
CAS Number: 1783561-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR