5-Ethyl-1,3-benzodioxole ≥95%

Supplier: AOBChem USA

Warning

222061-1G 222061-500MG 222061-5G 222061-25G
77466-998EA 341.96 USD
77466-998 77479-726 77490-020 77500-266
5-Ethyl-1,3-benzodioxole ≥95%
5-Ethyl-1,3-benzodioxole

Precaución: For research use only

Formula: C₉H₁₀O₂
Storage Temperature: Ambient
CAS Number: 60373-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR