4-Fluoro-6-methoxy-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

22105-1G 22105-500MG 22105-5G 22105-25G
77466-994EA 191.58 USD
77466-994 77479-722 77490-016 77500-264
4-Fluoro-6-methoxy-1H-indazole ≥95%
4-Fluoro-6-methoxy-1H-indazole

Precaución: For research use only

Formula: C₈H₇FN₂O
Storage Temperature: Ambient
MDL Number: MFCD07781917
CAS Number: 887569-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR