1-(2-Chloropyridin-3-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

220222-1G 220222-500MG 220222-5G 220222-25G
77466-986EA 268.83 USD
77466-986 77479-714 77490-008 77500-258
1-(2-Chloropyridin-3-yl)propan-1-one ≥95%
1-(2-Chloropyridin-3-yl)propan-1-one

Precaución: For research use only

Formula: C₈H₈ClNO
MW: 169.6082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847249
CAS Number: 131109-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR