1-(6-Bromo-2-chloropyridin-3-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

220140-1G 220140-500MG 220140-5G 220140-25G
77466-982EA 415.09 USD
77466-982 77479-710 77490-004 77500-254
1-(6-Bromo-2-chloropyridin-3-yl)propan-1-one ≥95%
1-(6-Bromo-2-chloropyridin-3-yl)propan-1-one

Precaución: For research use only

Formula: C₈H₇BrClNO
MW: 248.5042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34869053
CAS Number: 2969383-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR