1-Propanone, 1-(4,6-dichloro-3-pyridinyl)- ≥96%

Supplier: AOBChem USA

Warning

219995-1G 219995-500MG 219995-5G 219995-25G
77466-978EA 260.59 USD
77466-978 77479-706 77489-998 77500-250
1-Propanone, 1-(4,6-dichloro-3-pyridinyl)- ≥96%
1-Propanone, 1-(4,6-dichloro-3-pyridinyl)-

Precaución: For research use only

Formula: C₈H₇Cl₂NO
MW: 204.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32670636
CAS Number: 166525-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR