1-Bromo-3-((4-methoxybenzyl)oxy)-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

21954-1G 21954-500MG 21954-5G 21954-25G
77466-974EA 267.8 USD
77466-974 77479-702 77489-994 77500-244
1-Bromo-3-((4-methoxybenzyl)oxy)-5-(trifluoromethyl)benzene ≥95%
1-Bromo-3-((4-methoxybenzyl)oxy)-5-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₅H₁₂BrF₃O₂
MW: 361.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043992
CAS Number: 1820715-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR