(3-Chloro-2-fluorophenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

219295-1G 219295-500MG 219295-5G
77466-970EA 834.3 USD
77466-970 77479-698 77489-988
(3-Chloro-2-fluorophenyl)cyclopropylmethanone ≥95%
(3-Chloro-2-fluorophenyl)cyclopropylmethanone

Precaución: For research use only

Formula: C₁₀H₈ClFO
MW: 198.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20400927
CAS Number: 1342111-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR