1-(2-Fluoro-3-methylphenyl)-1-propanone ≥96%

Supplier: AOBChem USA

Warning

219157-1G 219157-500MG 219157-5G 219157-25G
77466-966EA 382.13 USD
77466-966 77479-694 77489-984 77500-240
1-(2-Fluoro-3-methylphenyl)-1-propanone ≥96%
1-(2-Fluoro-3-methylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₁₁FO
MW: 166.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397080
CAS Number: 1261675-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR