2-Bromo-4-(tert-pentyl)phenol ≥95%

Supplier: AOBChem USA

Warning

219062-1G 219062-500MG 219062-5G 219062-25G
77466-962EA 173.04 USD
77466-962 77479-690 77489-980 77500-234
2-Bromo-4-(tert-pentyl)phenol ≥95%
2-Bromo-4-(tert-pentyl)phenol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD09055133
CAS Number: 18966-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR