2,3-Dibromo-4-fluorobenzaldehyde ≥96%

Supplier: AOBChem USA

Warning

218801-1G 218801-500MG 218801-5G 218801-25G
77466-958EA 203.94 USD
77466-958 77479-686 77489-976 77500-232
2,3-Dibromo-4-fluorobenzaldehyde ≥96%
2,3-Dibromo-4-fluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₃Br₂FO
MW: 281.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784585
CAS Number: 1806327-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR