1,4-Dimethoxy-2-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

218434-1G 218434-500MG 218434-5G 218434-25G
77466-954EA 415.09 USD
77466-954 77479-682 77489-972 77500-228
1,4-Dimethoxy-2-[(1-methylethyl)thio]benzene ≥95%
1,4-Dimethoxy-2-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₁H₁₆O₂S
MW: 212.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030439
CAS Number: 51506-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR