3-Bromo-2,5-dimethoxypyridine ≥95%

Supplier: AOBChem USA

Warning

21574-1G 21574-500MG 21574-5G 21574-25G
77466-942EA 372.86 USD
77466-942 77479-672 77489-960 77500-214
3-Bromo-2,5-dimethoxypyridine ≥95%
3-Bromo-2,5-dimethoxypyridine

Precaución: For research use only

Formula: C₇H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD26398166
CAS Number: 1093951-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR