5-Fluoro-2-(trifluoromethyl)-4-pyridinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

215700-1G 215700-500MG 215700-5G
77466-938EA 889.92 USD
77466-938 77479-668 77489-956
5-Fluoro-2-(trifluoromethyl)-4-pyridinecarboxylic acid ≥97%
5-Fluoro-2-(trifluoromethyl)-4-pyridinecarboxylic acid

Precaución: For research use only

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256606
CAS Number: 1256806-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR