2,6-Difluoro-3-(phenylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21521-1G 21521-5G 21521-25G
77466-934EA 64.89 USD
77466-934 77489-952 77500-208
2,6-Difluoro-3-(phenylmethoxy)benzaldehyde ≥97%
2,6-Difluoro-3-(phenylmethoxy)benzaldehyde

Precaución: For research use only

Formula: C₁₄H₁₀F₂O₂
MW: 248.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527960
CAS Number: 152434-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR