α-Butyl-4-methoxy-3,5-dimethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

214641-1G 214641-500MG 214641-5G 214641-25G
77466-930EA 369.77 USD
77466-930 77479-662 77489-948 77500-204
α-Butyl-4-methoxy-3,5-dimethylbenzenemethanol ≥97%
α-Butyl-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24363305
CAS Number: 1548870-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR