α-Butyl-3-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214562-1G 214562-500MG 214562-5G
77466-922EA 546.93 USD
77466-922 77479-654 77489-940
α-Butyl-3-(methylthio)benzenemethanol ≥95%
α-Butyl-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867666
CAS Number: 2151201-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR