α-Butyl-2,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214535-1G 214535-500MG 214535-5G
77466-918EA 471.74 USD
77466-918 77479-650 77489-936
α-Butyl-2,5-dimethylbenzenemethanol ≥95%
α-Butyl-2,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD12787266
CAS Number: 854169-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR