α-(1-Methylpropyl)-4-(2-Methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214458-1G 214458-500MG 214458-5G 214458-25G
77466-914EA 415.09 USD
77466-914 77479-646 77489-932 77500-192
α-(1-Methylpropyl)-4-(2-Methylpropyl)benzenemethanol ≥95%
α-(1-Methylpropyl)-4-(2-Methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786723
CAS Number: 1248305-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR