2,6-Difluoro-4-methoxy-α-(1-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214453-1G 214453-500MG 214453-5G 214453-25G
77466-910EA 415.09 USD
77466-910 77479-642 77489-928 77500-188
2,6-Difluoro-4-methoxy-α-(1-methylpropyl)benzenemethanol ≥95%
2,6-Difluoro-4-methoxy-α-(1-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆F₂O₂
MW: 230.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786568
CAS Number: 1249070-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR